CAS RN 194611-64-8
IUPAC Name:
gallium(3+) tris(diethylazanide)
Synonyms:
gallium(3+) tris(diethylazanide)
Molecular Weight (g/mol):
286.12
Molecular Formula:
C12H30GaN3
InChi Key:
UZYORTJMAUFROH-UHFFFAOYSA-N
SMILES:
[Ga+3].CC[N-]CC.CC[N-]CC.CC[N-]CC
1
–
1
of
1
results
1
Tris(diethylamino)gallium(III), 99.9% (metals basis), Thermo Scientific™
CAS: 194611-64-8 Molecular Formula: C12H30GaN3 Molecular Weight (g/mol): 286.116 MDL Number: MFCD01863475 InChI Key: UZYORTJMAUFROH-UHFFFAOYSA-N Synonym: tris diethylamino gallium iii PubChem CID: 134068145 IUPAC Name: gallium;diethylazanide SMILES: CC[N-]CC.CC[N-]CC.CC[N-]CC.[Ga+3]