accessibility menu, dialog, popup

UserName

CAS RN 193-39-5

IUPAC Name: hexacyclo[16.3.1.0²,⁷.0⁸,²¹.0¹¹,²⁰.0¹⁴,¹⁹]docosa-1(21),2,4,6,8,10,12,14,16,18(22),19-undecaene
Synonyms: indeno(1,2,3-CD)pyrene
Molecular Weight (g/mol): 276.34
Molecular Formula: C22H12
InChi Key: SXQBHARYMNFBPS-UHFFFAOYSA-N
SMILES: C1=CC=C2C(=C1)C1=CC=C3C=CC4=CC=CC5=CC2=C1C3=C45
Molecular Weight (g/mol) 
  • (3)
  • (7)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)
  • (1)
Concentration 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (2)
  • (3)
  • (7)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)