CAS RN 19295-81-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Heptyl Nonyl Phthalate, TRC
CAS: 19295-81-9 Molecular Formula: C24H38O4 Molecular Weight (g/mol): 390.56 Synonym: 1,2-Benzenedicarboxylic Acid, Heptyl Nonyl Ester,Phthalic Acid Heptyl Nonyl Ester,1,2-Benzenedicarboxylic Acid, 1-Heptyl 2-Nonyl Ester SMILES: O=C(OCCCCCCCCC)C1=CC=CC=C1C(OCCCCCCC)=O