accessibility menu, dialog, popup

UserName

BOOH

CAS RN 19206-51-0

IUPAC Name: 3,4-dihydro-1H-2,1-benzoxaborinin-1-ol
Synonyms: 3,4-dihydro-2,1-benzoxaborinin-1-ol
Molecular Weight (g/mol): 147.97
Molecular Formula: C8H9BO2
InChi Key: CKEWCFGVNDVQSC-UHFFFAOYSA-N
SMILES: OB1OCCC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results