CAS RN 19186-97-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
tris(1,1,3-Tribromo-2,2-dimethylpropyl)phosphate, TRC
CAS: 19186-97-1 Molecular Formula: C15 H24 Br9 O4 P Molecular Weight (g/mol): 1018.46 Synonym: 1-Propanol, 3-bromo-2,2-bis(bromomethyl)-, 1,1',1' '-phosphate,1-Propanol, 3-bromo-2,2-bis(bromomethyl)-, phosphate (3:1) (8CI,9CI),CR 900,FR 370,FR 372,Flame Cut 175,Flame Cut 175R,Kronitex PB 370,PB 370,Reoflam FR 370,TPB 3070,TexFRon 3000,Tris(tribromoneopentyl) phosphate,Tris[2,2-bis(bromomethyl)-3-bromopropyl] phosphate,Tris[3-bromo-2,2-bis(bromomethyl)propyl] phosphate IUPAC Name: tris[3-bromo-2,2-bis(bromomethyl)propyl] phosphate SMILES: BrCC(CBr)(CBr)COP(=O)(OCC(CBr)(CBr)CBr)OCC(CBr)(CBr)CBr