accessibility menu, dialog, popup

UserName

H3COOCH3SH2NOO

CAS RN 19116-90-6

IUPAC Name: 2,5-dimethoxybenzene-1-sulfonamide
Synonyms: 2,5-dimethoxybenzenesulfonamide
Molecular Weight (g/mol): 217.24
Molecular Formula: C8H11NO4S
InChi Key: MMHMYFWOECSGDR-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(=C1)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results