CAS RN 19094-75-8
CAS RN 19094-75-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(2-Chloro-6-methylphenoxy)acetic Acid, TRC
CAS: 19094-75-8 Molecular Formula: C9H9O3Cl Molecular Weight (g/mol): 200.61 Synonym: 2-Chloro-6-methylphenoxyacetic Acid,2-(2-Chloro-6-methylphenoxy)acetic acid (ACI),Acetic acid, (2-chloro-6-methylphenoxy)- (9CI),Acetic acid, [(6-chloro-o-tolyl)oxy]- (8CI),(2-Chloro-6-methylphenoxy)acetic acid,(2-Methyl-6-chlorophenoxy)acetic acid,(6-Chloro-2-methylphenoxy)acetic acid,NSC 190539 IUPAC Name: 2-(2-chloro-6-methylphenoxy)acetic acid SMILES: Cc1cccc(Cl)c1OCC(=O)O