accessibility menu, dialog, popup

UserName

NOHICl

CAS RN 188057-26-3

IUPAC Name: 6-chloro-2-iodopyridin-3-ol
Synonyms: 6-chloro-2-iodopyridin-3-ol
Molecular Weight (g/mol): 255.44
Molecular Formula: C5H3ClINO
InChi Key: FWIMPBYPMQSSCD-UHFFFAOYSA-N
SMILES: OC1=C(I)N=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results