accessibility menu, dialog, popup

UserName

CAS RN 186517-29-3

CH3H3CCH3ClFO

CAS RN 186517-29-3

IUPAC Name: 6-tert-butyl-3-chloro-2-fluorobenzaldehyde
Synonyms: 6-tert-butyl-3-chloro-2-fluorobenzaldehyde
Molecular Weight (g/mol): 214.66
Molecular Formula: C11H12ClFO
InChi Key: YHMGAQILYKLARE-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(Cl)C(F)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results