accessibility menu, dialog, popup

UserName

CAS RN 18527-19-0

ClSN

CAS RN 18527-19-0

IUPAC Name: 2-[(4-chlorophenyl)sulfanyl]acetonitrile
Synonyms: 2-[(4-chlorophenyl)sulfanyl]acetonitrile
Molecular Weight (g/mol): 183.65
Molecular Formula: C8H6ClNS
InChi Key: WZXBUVDTYAPXOQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(SCC#N)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results