CAS RN 182219-43-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(4’-Hydroxyphenyl)-2-butanone-d5, TRC
CAS: 182219-43-8 Molecular Formula: C10H7D5O2 Molecular Weight (g/mol): 169.23 Synonym: 2-Butanone-1,1,1,3,3-d5, 4-(4-hydroxyphenyl)- (9CI),1,1,1,3,3-Pentadeuterio-4-(4-hydroxyphenyl)butan-2-one,4-(4-Hydroxyphenyl)butan-2-one-1,1,1,3,3-D5,(p-Hydroxybenzyl)acetone-d5,1-(p-Hydroxyphenyl)-3-butanone-d5,Frambinone-d5,Oxyphenylon-d5,Raspberry ketone-d5,Rheosmin-d5 IUPAC Name: 1,1,1,3,3-pentadeuterio-4-(4-hydroxyphenyl)butan-2-one SMILES: [2H]C([2H])([2H])C(=O)C([2H])([2H])Cc1ccc(O)cc1