accessibility menu, dialog, popup

UserName

LiH3CSiCH3CH3CH2

CAS RN 1822-00-0

IUPAC Name: lithium(1+) (trimethylsilyl)methanide
Synonyms: lithium(1+) (trimethylsilyl)methanide
Molecular Weight (g/mol): 94.16
Molecular Formula: C4H11LiSi
InChi Key: KVWLUDFGXDFFON-UHFFFAOYSA-N
SMILES: [Li+].C[Si](C)(C)[CH2-]
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)