CAS RN 1821143-80-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
DMU2139, MedChemExpress
MedChemExpress DMU2139 is a potent and specific CYP1B1 inhibitor, with IC50s of 9 nM and 795 nM for CYP1B1 and CYP1A1, respectively.