CAS RN 180918-40-5
IUPAC Name:
1-(trifluoromethyl)cyclohexane-1-carboxylic acid
Synonyms:
1-(trifluoromethyl)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
196.17
Molecular Formula:
C8H11F3O2
InChi Key:
VFSFTDFBHSFDEE-UHFFFAOYSA-N
SMILES:
OC(=O)C1(CCCCC1)C(F)(F)F