accessibility menu, dialog, popup

UserName

CAS RN 180608-37-1

IUPAC Name: 2-bromo-6-fluoro-4-methylpyridine
Synonyms: 2-bromo-6-fluoro-4-methylpyridine
Molecular Weight (g/mol): 190.02
Molecular Formula: C6H5BrFN
InChi Key: CIDWWTCSJWQJQR-UHFFFAOYSA-N
SMILES: CC1=CC(F)=NC(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results