CAS RN 1799948-06-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
HIF-2α-IN-1, MedChemExpress
MedChemExpress HIF-2(alpha)-IN-1 is a HIF-2(alpha) inhibitor has an IC50 of less than 500 nM in HIF-2(alpha) scintillation proximity assay.