CAS RN 17978-75-5
IUPAC Name:
tris(1,1,1,2,2,3,3-heptafluoro-6-hydroxy-7,7-dimethyloct-5-en-4-one) erbium
Synonyms:
tris(1,1,1,2,2,3,3-heptafluoro-6-hydroxy-7,7-dimethyloct-5-en-4-one) erbium
Molecular Weight (g/mol):
1055.81
Molecular Formula:
C30H33ErF21O6
InChi Key:
LEHFMLQRGJCLOZ-UHFFFAOYSA-N
SMILES:
[Er].CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(O)=CC(=O)C(F)(F)C(F)(F)C(F)(F)F
1
–
1
of
1
results
1
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)erbium(III), 97+%, Thermo Scientific™
CAS: 17978-75-5 Molecular Formula: C30H33ErF21O6 Molecular Weight (g/mol): 1055.81 MDL Number: MFCD00074990 InChI Key: LEHFMLQRGJCLOZ-UHFFFAOYSA-N Synonym: resolve-al? erfod