CAS RN 1795031-70-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diethylcarbamazine-d3 Citrate, TRC
CAS: 1795031-70-7 Molecular Formula: C16H26D3N3O8 Molecular Weight (g/mol): 394.44 Synonym: N,N-Diethyl-4-(methyl-d3)-1-piperazinecarboxamide Citrate Salt,N,N-Diethyl-4-(methyl-d3)-1-piperazinecarboxamide 2-Hydroxy-1,2,3-propanetricarboxylate,Dirocide-d3,Ditrazin-d3 Citrate,Ditrazine-d3,Ethodryl-d3 Citrate,Ethylaminoazine-d3 Citrate,Filaribits-d3,Filazine-d3,Franocide-d3,Franozan-d3,Hetrazan-d3,Longicid,-d3 Loxuran-d3,N,N-Diethyl-4-(methyl-d3)-1-piperazinecarboxamide Dihydrogen Citrate,NSC 80513-d3 SMILES: O=C(N(CC)CC)N1CCN(C([2H])([2H])[2H])CC1.OC(C[C](O)=O)(C(O)=O)CC(O)=O