CAS RN 1794759-59-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Hydroxy Carbofuran-d3, TRC
CAS: 1794759-59-3 Molecular Formula: C12H12D3NO4 Molecular Weight (g/mol): 240.27 Synonym: 2,3-Dihydro-2,2-dimethyl-3,7-benzofurandiol 7-(N-Methylcarbamate-d3),(Methyl-d3)carbamic Acid 2,3-Dihydro-3-hydroxy-2,2-dimethyl-7-benzofuranyl Ester,2,3-Dihydro-2,2-dimethyl-3-hydroxy-7-benzofuranyl N-(Methyl-d3)carbamate,2,3-Dihydro-2,2-dimethyl-3-hydroxy-7-benzofuranyl (Methyl-d3)carbamate,2,3-Dihydro-3-hydroxy-2,2-dimethyl-7-benzofuranyl (Methyl-d3)carbamate SMILES: O=C(NC([2H])([2H])[2H])OC1=C2C(C(O)C(C)(C)O2)=CC=C1