accessibility menu, dialog, popup

UserName

CAS RN 1779-10-8

IUPAC Name: 4,7-dibromo-3-hydroxynaphthalene-2-carboxylic acid
Synonyms: 4,7-dibromo-3-hydroxynaphthalene-2-carboxylic acid
Molecular Weight (g/mol): 345.97
Molecular Formula: C11H6Br2O3
InChi Key: WNMKUIQCIRAXBN-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(O)C(Br)=C2C=CC(Br)=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results