CAS RN 17551-52-9
IUPAC Name:
6-chloro-2-methyl-1,4-dihydropyrimidin-4-one
Synonyms:
6-chloro-2-methyl-1H-pyrimidin-4-one
Molecular Weight (g/mol):
144.56
Molecular Formula:
C5H5ClN2O
InChi Key:
QNANRGHPXMUJQC-UHFFFAOYSA-N
SMILES:
CC1=NC(=O)C=C(Cl)N1
1
–
1
of
1
results
1
6-Chloro-4-hydroxy-2-methylpyrimidine, 97%
CAS: 17551-52-9 Molecular Formula: C5H5ClN2O Molecular Weight (g/mol): 144.558 MDL Number: MFCD00030773 InChI Key: QNANRGHPXMUJQC-UHFFFAOYSA-N Synonym: 6-chloro-2-methyl-4-pyrimidinol,6-chloro-2-methylpyrimidin-4-ol,6-chloro-2-methylpyrimidin-4 1h-one,4 1h-pyrimidinone, 6-chloro-2-methyl,chloromethylpyrimidinol,4 3h-pyrimidinone, 6-chloro-2-methyl,4-pyrimidinol, 6-chloro-2-methyl,6-chloro-2-methyl-4 3h-pyrimidone,2-methyl-4-hydroxy-6-chloropyrimidine,4-chloro-6-hydroxy-2-methylpyrimidine PubChem CID: 295975 IUPAC Name: 6-chloro-2-methyl-1H-pyrimidin-4-one SMILES: CC1=NC(=O)C=C(N1)Cl