accessibility menu, dialog, popup

UserName

NOOHOOO

CAS RN 174800-02-3

IUPAC Name: 4-(benzyloxy)-1-{[(9H-fluoren-9-yl)methoxy]carbonyl}pyrrolidine-2-carboxylic acid
Synonyms: 4-(benzyloxy)-1-[(9H-fluoren-9-ylmethoxy)carbonyl]pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 443.5
Molecular Formula: C27H25NO5
InChi Key: XGFMHBUVVWZBFT-UHFFFAOYNA-N
SMILES: OC(=O)C1CC(CN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results