accessibility menu, dialog, popup

UserName

CAS RN 174732-59-3

IUPAC Name: 2-amino-3-(3-fluoro-4-methylphenyl)propanoic acid
Synonyms: 2-amino-3-(3-fluoro-4-methylphenyl)propanoic acid
Molecular Weight (g/mol): 197.21
Molecular Formula: C10H12FNO2
InChi Key: MUTNDIMQYVVGMX-UHFFFAOYNA-N
SMILES: CC1=C(F)C=C(CC(N)C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results