CAS RN 1733-89-7
CAS RN 1733-89-7
IUPAC Name:
potassium naphthalen-2-yl sulfate
Synonyms:
potassium naphthalen-2-yl sulfate
Molecular Weight (g/mol):
262.32
Molecular Formula:
C10H7KO4S
InChi Key:
SKROFUUKJBLLKJ-UHFFFAOYSA-M
SMILES:
[K+].[O-]S(=O)(=O)OC1=CC=C2C=CC=CC2=C1
1
–
1
of
1
results
1
2-Naphthyl sulfate potassium salt, 98%
CAS: 1733-89-7 Molecular Formula: C10H8KO4S Molecular Weight (g/mol): 263.328 MDL Number: MFCD00004059 InChI Key: KPQVGLIXPMOHMI-UHFFFAOYSA-N Synonym: 2-naphthyl hydrogen sulfate PubChem CID: 24186395 IUPAC Name: naphthalen-2-yl hydrogen sulfate;potassium SMILES: C1=CC=C2C=C(C=CC2=C1)OS(=O)(=O)O.[K]