CAS RN 173200-56-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(E)-6,6-Dimethylhept-2-en-4-yn-1-ol, TRC
CAS: 173200-56-1 Molecular Formula: C9H14O Molecular Weight (g/mol): 138.21 Synonym: (2E)-6,6-Dimethyl-2-hepten-4-yn-1-ol IUPAC Name: (E)-6,6-dimethylhept-2-en-4-yn-1-ol SMILES: CC(C)(C)C#C\C=C\CO