CAS RN 17250-25-8
IUPAC Name:
iridium(1+) methanediidylidyneoxidanium tris(triphenylphosphane) hydride
Synonyms:
iridium(1+) methanediidylidyneoxidanium tris(triphenylphosphine) hydride
Molecular Weight (g/mol):
1008.11
Molecular Formula:
C55H46IrOP3
InChi Key:
ZCRCZIFIPMYSDK-UHFFFAOYSA-N
SMILES:
[H-].[Ir+].[C--]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Carbonylhydridotris(triphenylphosphine)iridium(I), Ir 18.6% min, Thermo Scientific™
CAS: 17250-25-8 Molecular Formula: C55H46IrOP3 Molecular Weight (g/mol): 1008.11 MDL Number: MFCD00015525 InChI Key: IOPVMNYABUEALT-UHFFFAOYSA-N Synonym: carbonylhydridotris triphenylphosphine iridium i,iridium 1+ formaldehyde tris triphenylphosphine hydride PubChem CID: 92135237 IUPAC Name: formaldehyde;hydride;iridium;triphenylphosphane SMILES: [IrH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1