CAS RN 17185-29-4
CAS RN 17185-29-4
IUPAC Name:
λ¹-rhodium(1+) methanediidylidyneoxidanium tris(triphenylphosphane) hydride
Synonyms:
λ¹-rhodium(1+) methanediidylidyneoxidanium tris(triphenylphosphine) hydride
Molecular Weight (g/mol):
918.8
Molecular Formula:
C55H46OP3Rh
InChi Key:
FLGOKRAQOPBGLK-UHFFFAOYSA-N
SMILES:
[H-].[Rh+].[C--]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Carbonylhydridotris(triphenylphosphine)rhodium(I), Rh 10.0% min
CAS: 17185-29-4 Molecular Formula: C55H46OP3Rh Molecular Weight (g/mol): 918.80 MDL Number: MFCD00151644 InChI Key: AACIZACVKFEETJ-UHFFFAOYSA-N Synonym: tristriphenylphosphine rhodium carbonyl hydride,hydridocarbonyltris triphenylphosphine rhodium i PubChem CID: 132451836 IUPAC Name: carbon monoxide;rhodium;triphenylphosphane SMILES: [RhH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1