accessibility menu, dialog, popup

UserName

FFFFClO

CAS RN 171243-30-4

IUPAC Name: 3-fluoro-5-(trifluoromethyl)benzoyl chloride
Synonyms: 3-fluoro-5-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 226.55
Molecular Formula: C8H3ClF4O
InChi Key: BGAKSLBTFLVNAH-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(=C1)C(F)(F)F)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results