CAS RN 17120-26-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-formyl-3,5-dimethyl-1H-pyrrole-2-carboxylic acid, TRC
CAS: 17120-26-2 Molecular Formula: C8H9NO3 Molecular Weight (g/mol): 167.16 SMILES: Cc1[nH]c(C(=O)O)c(C)c1C=O