accessibility menu, dialog, popup

UserName

CAS RN 170886-13-2

NHOFFF

CAS RN 170886-13-2

IUPAC Name: 2-(trifluoromethyl)-1,4-dihydropyridin-4-one
Synonyms: 2-(trifluoromethyl)-1H-pyridin-4-one
Molecular Weight (g/mol): 163.1
Molecular Formula: C6H4F3NO
InChi Key: LKHCWMQMRXAPHX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=O)C=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results