accessibility menu, dialog, popup

UserName

CAS RN 169760-16-1

CH3OHNBOHHO

CAS RN 169760-16-1

IUPAC Name: (2-acetamidophenyl)boronic acid
Synonyms: 2-acetamidophenylboronic acid
Molecular Weight (g/mol): 178.98
Molecular Formula: C8H10BNO3
InChi Key: UMOPBIVXPOETPG-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results