CAS RN 169530-97-6
IUPAC Name:
2-amino-3-(3-nitrophenyl)propanoic acid
Synonyms:
2-amino-3-(3-nitrophenyl)propanoic acid
Molecular Weight (g/mol):
210.19
Molecular Formula:
C9H10N2O4
InChi Key:
YTHDRUZHNYKZGF-UHFFFAOYNA-N
SMILES:
NC(CC1=CC(=CC=C1)[N+]([O-])=O)C(O)=O
1
–
1
of
1
results
1
3-Nitro-D-phenylalanine, 95%
CAS: 169530-97-6 Molecular Formula: C9H10N2O4 Molecular Weight (g/mol): 210.19 MDL Number: MFCD04117829 InChI Key: YTHDRUZHNYKZGF-UHFFFAOYNA-N Synonym: r-2-amino-3-3-nitrophenyl propanoic acid,3-nitro-d-phenylalanine,d-3-nitrophenylalanine,2r-2-amino-3-3-nitrophenyl propanoic acid,d-3-no2-phe-oh,d-3-nitrophe,d-3-nitro phenylalanine,r-2-amino-3-3-nitrophenyl propanoicacid PubChem CID: 7000156 IUPAC Name: 2-amino-3-(3-nitrophenyl)propanoic acid SMILES: NC(CC1=CC(=CC=C1)[N+]([O-])=O)C(O)=O