CAS RN 1689566-03-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Flupyrimin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Flupyrimin, TRC
CAS: 1689566-03-7 Molecular Formula: C13 H9 Cl F3 N3 O Molecular Weight (g/mol): 315.68 Synonym: Acetamide, N-[1-[(6-chloro-3-pyridinyl)methyl]-2(1H)-pyridinylidene]-2,2,2-trifluoro-, [N(E)]-,[N(E)]-N-[1-[(6-Chloro-3-pyridinyl)methyl]-2(1H)-pyridinylidene]-2,2,2-trifluoroacetamide,Flupyrimin,ME 5382 IUPAC Name: (NE)-N-[1-[(6-chloropyridin-3-yl)methyl]pyridin-2-ylidene]-2,2,2-trifluoroacetamide SMILES: FC(F)(F)C(=O)\N=C\1/C=CC=CN1Cc2ccc(Cl)nc2