CAS RN 16851-82-4
CAS RN 16851-82-4
IUPAC Name:
1-(benzenesulfonyl)-1H-pyrrole
Synonyms:
1-(benzenesulfonyl)pyrrole
Molecular Weight (g/mol):
207.25
Molecular Formula:
C10H9NO2S
InChi Key:
PPPXRIUHKCOOMU-UHFFFAOYSA-N
SMILES:
O=S(=O)(N1C=CC=C1)C1=CC=CC=C1
1
–
3
of
3
results
2
1-(Phenylsulfonyl)pyrrole, 98%
CAS: 16851-82-4 Molecular Formula: C10H9NO2S Molecular Weight (g/mol): 207.25 MDL Number: MFCD00067739 InChI Key: PPPXRIUHKCOOMU-UHFFFAOYSA-N PubChem CID: 140146 IUPAC Name: 1-(benzenesulfonyl)pyrrole SMILES: O=S(=O)(N1C=CC=C1)C1=CC=CC=C1