accessibility menu, dialog, popup

UserName

CAS RN 16832-21-6

IUPAC Name: (1R)-1-[(3'aR,5'R,6'S,6'aR)-6'-hydroxy-tetrahydrospiro[cyclohexane-1,2'-furo[2,3-d][1,3]dioxol]-5'-yl]ethane-1,2-diol
Synonyms: (1R)-1-[(3'aR,5'R,6'S,6'aR)-6'-hydroxy-tetrahydrospiro[cyclohexane-1,2'-furo[2,3-d][1,3]dioxol]-5'-yl]ethane-1,2-diol
Molecular Weight (g/mol): 260.29
Molecular Formula: C12H20O6
InChi Key: KAUCUKASFLXURN-RCZSTQMZSA-N
SMILES: OC[C@@H](O)[C@H]1O[C@@H]2OC3(CCCCC3)O[C@@H]2[C@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results