CAS RN 16827-96-6
CAS RN 16827-96-6
IUPAC Name:
λ²-cobalt(2+) ammonium hydrate phosphate
Synonyms:
λ²-cobalt(2+) ammonium hydrate phosphate
Molecular Weight (g/mol):
189.96
Molecular Formula:
CoH6NO5P
InChi Key:
PNSIYUITZYKGRY-UHFFFAOYSA-L
SMILES:
[NH4+].O.[Co++].[O-]P([O-])([O-])=O