accessibility menu, dialog, popup

UserName

CH3OOOCH3OCH3

CAS RN 16740-98-0

IUPAC Name: 3,4-dimethoxy-2-(methoxymethyl)-3,4-dihydro-2H-pyran
Synonyms: 3,4-dimethoxy-2-(methoxymethyl)-3,4-dihydro-2H-pyran
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: PZJPNKKRMNFDQJ-UHFFFAOYNA-N
SMILES: COCC1OC=CC(OC)C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results