CAS RN 16732-65-3
CAS RN 16732-65-3
IUPAC Name:
6-bromo-1H-indole-2-carboxylic acid
Synonyms:
6-bromo-1H-indole-2-carboxylic acid
Molecular Weight (g/mol):
240.06
Molecular Formula:
C9H6BrNO2
InChi Key:
SVBVYRYROZWKNJ-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC2=CC=C(Br)C=C2N1
1
–
1
of
1
results
1
6-Bromoindole-2-carboxylic acid, 97%
CAS: 16732-65-3 Molecular Formula: C9H6BrNO2 Molecular Weight (g/mol): 240.056 MDL Number: MFCD02664469 InChI Key: SVBVYRYROZWKNJ-UHFFFAOYSA-N Synonym: 6-bromoindole-2-carboxylic acid,6-bromo-2-indolecarboxylic acid,1h-indole-2-carboxylic acid, 6-bromo,6-bromoindole-2-carboxylicacid,zlchem 475,pubchem1664,acmc-1bqkh,intermediates-zcf02255,evoblocks eb21020,timtec-bb sbb014280 PubChem CID: 4011696 IUPAC Name: 6-bromo-1H-indole-2-carboxylic acid SMILES: C1=CC2=C(C=C1Br)NC(=C2)C(=O)O