accessibility menu, dialog, popup

UserName

CAS RN 16732-60-8

HNOHONOO

CAS RN 16732-60-8

IUPAC Name: 4-nitro-1H-indole-2-carboxylic acid
Synonyms: 4-nitro-1H-indole-2-carboxylic acid
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H6N2O4
InChi Key: MDWCXENHATUMDQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=C(C=CC=C2N1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results