accessibility menu, dialog, popup

UserName

CAS RN 166328-08-1

IUPAC Name: potassium trifluoro[4-(trifluoromethyl)phenyl]boranuide
Synonyms: potassium trifluoro[4-(trifluoromethyl)phenyl]boranuide
Molecular Weight (g/mol): 252.01
Molecular Formula: C7H4BF6K
InChi Key: KVDSXGFQLXKOID-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results