accessibility menu, dialog, popup

UserName

CAS RN 164171-56-6

CH3HNOO(E/Z)

CAS RN 164171-56-6

IUPAC Name: 6-(2,5-dihydro-1,2-oxazol-5-ylidene)-4-methylcyclohexa-2,4-dien-1-one
Synonyms: 4-methyl-6-(2H-1,2-oxazol-5-ylidene)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: CDUCCINDCURBGI-UHFFFAOYSA-N
SMILES: CC1=CC(=C2ONC=C2)C(=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results