CAS RN 1639417-53-0
CAS RN 1639417-53-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tenalisib, MedChemExpress
MedChemExpress Tenalisib (RP6530) is a novel, potent, and selective PI3Kδ and PI3Kγ inhibitor with IC50 values of 25 and 33 nM, respectively.