CAS RN 162849-95-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-[(1S,2S,4S)-4-[[(1,1-Dimethylethoxy)carbonyl]amino]-2-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]carbamic Acid 5-Thiazolylmethyl Ester, TRC
CAS: 162849-95-8 Molecular Formula: C28H35N3O5S Molecular Weight (g/mol): 525.66 Synonym: [1S-(1R*,2R*,4R*)]-[4-[[(1,1-Dimethylethoxy)carbonyl]amino]-2-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]carbamic Acid 5-Thiazolylmethyl Ester,tert-Butyl N-[(1S,3S,4S)-1-Benzyl-3-hydroxy-5-phenyl-4-[[(thiazol-5-ylmethoxy)carbonyl]amino]pentyl]carbamate,Ritonavir EP Impurity J SMILES: CC(C)(C)OC(=O)N[C@H](C[C@H](O)[C@H](Cc1ccccc1)NC(=O)OCc2cncs2)Cc3ccccc3