accessibility menu, dialog, popup

UserName

OH(S)OO

CAS RN 1621-55-2

IUPAC Name: (2S)-2-(3-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: (2S)-3'-hydroxyflavanone
Molecular Weight (g/mol): 240.26
Molecular Formula: C15H12O3
InChi Key: JVSPTYZZNUXJHN-HNNXBMFYSA-N
SMILES: OC1=CC=CC(=C1)[C@@H]1CC(=O)C2=CC=CC=C2O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results