accessibility menu, dialog, popup

UserName

NNN

CAS RN 160017-13-0

IUPAC Name: 6-(pyrrolidin-1-yl)pyridine-2-carbonitrile
Synonyms: 6-(pyrrolidin-1-yl)pyridine-2-carbonitrile
Molecular Weight (g/mol): 173.22
Molecular Formula: C10H11N3
InChi Key: KUVMTKFDWSRABZ-UHFFFAOYSA-N
SMILES: N#CC1=NC(=CC=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results