accessibility menu, dialog, popup

UserName

H3CNNCl

CAS RN 1578-97-8

IUPAC Name: 3-(chloromethyl)-1-methyl-1H-indazole
Synonyms: 3-(chloromethyl)-1-methylindazole
Molecular Weight (g/mol): 180.64
Molecular Formula: C9H9ClN2
InChi Key: LYLGQIOTMAQQEB-UHFFFAOYSA-N
SMILES: CN1N=C(CCl)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results