accessibility menu, dialog, popup

UserName

PPClClPtH3CH3CH3CH3CCH3H3C

CAS RN 15692-07-6

IUPAC Name: dichloroplatinum; bis(triethylphosphane)
Synonyms: platinum(II) chloride; bis(triethylphosphine)
Molecular Weight (g/mol): 502.3
Molecular Formula: C12H30Cl2P2Pt
InChi Key: WPWLTKRUFHHDLP-UHFFFAOYSA-L
SMILES: Cl[Pt]Cl.CCP(CC)CC.CCP(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results