CAS RN 15687-41-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Oxyfedrine, TRC
CAS: 15687-41-9 Molecular Formula: C19 H23 N O3 Molecular Weight (g/mol): 313.39 Synonym: 3-[[(1S,2R)-2-Hydroxy-1-methyl-2-phenylethyl]amino]-1-(3-methoxyphenyl)-1-propanone IUPAC Name: 3-[[(1R,2S)-1-hydroxy-1-phenylpropan-2-yl]amino]-1-(3-methoxyphenyl)propan-1-one SMILES: COc1cccc(c1)C(=O)CCN[C@@H](C)[C@H](O)c2ccccc2