accessibility menu, dialog, popup

UserName

NHNOI

CAS RN 155798-08-6

IUPAC Name: N-(1-benzylpiperidin-4-yl)-4-iodobenzamide
Synonyms: N-(1-benzylpiperidin-4-yl)-4-iodobenzamide
Molecular Weight (g/mol): 420.29
Molecular Formula: C19H21IN2O
InChi Key: HELCSESNNDZLFM-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=C1)C(=O)NC1CCN(CC2=CC=CC=C2)CC1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results