accessibility menu, dialog, popup

UserName

CAS RN 15480-00-9

HOOCl

CAS RN 15480-00-9

IUPAC Name: 2-(2-chlorophenoxy)ethan-1-ol
Synonyms: ETHYLENEGLYCOLMONO2chlorophenylether
Molecular Weight (g/mol): 172.61
Molecular Formula: C8H9ClO2
InChi Key: FDQGMCQSIVZGHW-UHFFFAOYSA-N
SMILES: OCCOC1=CC=CC=C1Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)